Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RNF190 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501988936
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-8936 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-8936 Supplier Bioss Supplier No. BS9245R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

MARCH10 is a member of the MARCH family of membrane-bound E3 ubiquitin ligases to target lysines in substrate proteins, thereby signaling their vesicular transport between membrane compartments .

Specifications

Antigen RNF190
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene 43900
Gene Accession No. Q8NA82
Gene Alias 10-Mar; 4933417C16Rik; AV039223; FLJ35757; March10; MARCH-X; membrane associated ring finger 10; membrane associated ring-CH-type finger 10; membrane-associated ring finger (C3HC4) 10; membrane-associated ring finger (C3HC4) 10, E3 ubiquitin protein ligase; Membrane-associated RING finger protein 10; Membrane-associated RING-CH protein X; microtubule-associated E3 ubiquitin ligase; OTTMUSG00000002847; Probable E3 ubiquitin-protein ligase MARCH10; RGD1311692; RING finger protein 190; RING-type E3 ubiquitin transferase MARCH10; RNF190; testis secretory sperm-binding protein Li 228n
Gene Symbols 43900
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human RNF190.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 162333
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.