Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RNF19A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA554861
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA554861 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA554861 Supplier Invitrogen™ Supplier No. PA554861
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: EMCTDKNSIF STNTSSDNGL TSISKQIGDF IECPLCLLRH SKDRFPDIMT CHHRSCVDCL Highest antigen sequence identity to the following orthologs: Mouse - 98%, Rat - 93%.

This gene encodes a member of the ring between ring fingers (RBR) protein family, and the encoded protein contains two RING-finger motifs and an in between RING fingers motif. This protein is an E3 ubiquitin ligase that is localized to Lewy bodies, and ubiquitylates synphilin-1, which is an interacting protein of alpha synuclein in neurons. The encoded protein may be involved in amyotrophic lateral sclerosis and Parkinson's disease. Alternative splicing results in multiple transcript variants. [provided by RefSeq, Jul 2013].
TRUSTED_SUSTAINABILITY

Specifications

Antigen RNF19A
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.20 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene RNF19A
Gene Accession No. Q9NV58
Gene Alias AA032313; Dorfin; Double ring-finger protein; E3 ubiquitin-protein ligase RNF19A; Gametogenesis-expressed protein GEG-154; Geg-154; p38; protein p38 interacting with transcription factor Sp1; ring finger protein (C3HC4 type) 19; ring finger protein 19; ring finger protein 19A; ring finger protein 19A, E3 ubiquitin protein ligase; ring finger protein 19A, RBR E3 ubiquitin protein ligase; ring-IBR-ring domain containing protein Dorfin; Rnf19; RNF19A; Ubce7ip2; UBCM4-interacting protein 117; ubiquitin conjugating enzyme 7 interacting protein 2; UIP117; XY body protein; XYbp
Gene Symbols RNF19A
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human RNF19A. Recombinant protein control fragment (Product #RP-93729).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 25897
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.