Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RNF19B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA562017
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA562017 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA562017 Supplier Invitrogen™ Supplier No. PA562017
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: AQPESIRSDL ESSDAQSDDV PDITSDECGS PRSHTAACPS TPRAQGAPSP SAHMNLSALA EGQTVLKPEG GEA Highest antigen sequence identity to the following orthologs: Mouse - 82%, Rat - 84%.

E3 ubiquitin-protein ligase which accepts ubiquitin from E2 ubiquitin-conjugating enzymes UBE2L3 and UBE2L6 in the form of a thioester and then directly transfers the ubiquitin to targeted substrates, such as UCKL1. Involved in the cytolytic activity of natural killer cells and cytotoxic T-cells.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RNF19B
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene RNF19B
Gene Accession No. Q6ZMZ0
Gene Alias 4930534K13Rik; 4930555L03Rik; E3 ubiquitin-protein ligase RNF19B; IBR domain containing 3; IBR domain-containing protein 3; IBRDC3; Natural killer lytic-associated molecule; NKLAM; RING finger protein 19B; Rnf19b
Gene Symbols RNF19B
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human RNF19B. Recombinant protein control fragment (Product #RP-101226).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 127544
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.