Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RNF2 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP188712
Description
RNF2 Polyclonal specifically detects RNF2 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
RNF2 | |
Polyclonal | |
Western Blot 0.04-0.4 ug/ml, Immunohistochemistry 1:50 - 1:200, Immunocytochemistry/Immunofluorescence 0.25-2 ug/ml, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
BAP-1, BAP1RING finger protein 1B, DINGRING finger protein BAP-1, EC 6.3.2, HIPI3RING1BE3 ubiquitin-protein ligase RING2, Huntingtin-interacting protein 2-interacting protein 3, Protein DinG, ring finger protein 2HIP2-interacting protein 3, RING1b, RING2EC 6.3.2.- | |
Rabbit | |
Affinity Purified | |
RUO | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Unconjugated | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
RNF2 | |
This antibody was developed against Recombinant Protein corresponding to amino acids:ALISKIYPSRDEYEAHQERVLARINKHNNQQALSHSIEEGLKIQAMNRLQRGKKQQIENGSGAEDNGDSSHCSNASTHSNQEAGPSNKRTKTSDDSGLELDNNNAAMAIDPVMDGASEIELV | |
0.1 mL | |
Apoptosis, Breast Cancer, Cancer, Signal Transduction, Transcription Factors and Regulators, Zinc Finger | |
6045 | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction