Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RNF2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA576758
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA576758 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA576758 Supplier Invitrogen™ Supplier No. PA576758
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).

RNF2 (E3 ubiquitin-protein ligase RING2) is a ligase that mediates monoubiquitination of Lys-119 of histone H2A (H2AK119Ub). Therefore, it plays a central role in histone code and gene regulation. The ligase activity is enhanced by BMI1/PCGF4. H2AK119Ub gives a specific tag for epigeneti transcriptional repression and participates in X chromosome inactivation of female mammals. RNF2 may be involved in the initiation of both imprinted and random X inactivation. RNF2 is also an essential component of a Polycomb group (PcG) multiprotein PRC1-like complex. This complex class is required to maintain the transcriptionally repressive state of many genes including Hox genes throughout development. The PcG PRC1 complex acts via chromatin remodeling and modification of histones and rendering chromatin heritably changed in its expressibility.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RNF2
Applications ChIP Assay, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.2
Gene RNF2
Gene Accession No. Q4KLY4, Q99496, Q9CQJ4
Gene Alias AI326319; AI450156; AU019207; BAP 1; BAP1; BAP-1; DinG; DinG protein; E3 ubiquitin-protein ligase RING2; fa19h05; GS1-120K12.1; HIP2-interacting protein 3; HIPI 3; HIPI3; Huntingtin-interacting protein 2-interacting protein 3; OTTHUMP00000060668; protein DinG; RING 1B; RING 2; RING finger protein 1B; ring finger protein 2; RING finger protein BAP-1; RING1B; RING2; RING-type E3 ubiquitin transferase RING2; RNF 2; Rnf2; wu:fa19h05; wz4556; wz4566
Gene Symbols RNF2
Host Species Rabbit
Immunogen Recombinant full length Human RNF2.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 19821, 304850, 6045
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.