Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RNF2 Rabbit anti-Human, Mouse, Rat, Clone: 9Q2W2, Novus Biologicals™

Rabbit Monoclonal Antibody
$205.50 - $458.50
Specifications
Antigen | RNF2 |
---|---|
Clone | 9Q2W2 |
Applications | Western Blot, Immunoprecipitation |
Classification | Monoclonal |
Conjugate | Unconjugated |
Description
RNF2 Monoclonal antibody specifically detects RNF2 in Human, Mouse, Rat samples. It is validated for Western Blot, ImmunoprecipitationSpecifications
RNF2 | |
Western Blot, Immunoprecipitation | |
Unconjugated | |
Rabbit | |
Apoptosis, Breast Cancer, Cancer, Signal Transduction, Transcription Factors and Regulators, Zinc Finger | |
PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
6045 | |
IgG | |
Affinity purified |
9Q2W2 | |
Monoclonal | |
Purified | |
RUO | |
Human, Mouse, Rat | |
BAP-1, BAP1RING finger protein 1B, DINGRING finger protein BAP-1, EC 6.3.2, HIPI3RING1BE3 ubiquitin-protein ligase RING2, Huntingtin-interacting protein 2-interacting protein 3, Protein DinG, ring finger protein 2HIP2-interacting protein 3, RING1b, RING2EC 6.3.2.- | |
A synthetic peptide corresponding to a sequence within amino acids 100-200 of human RNF2 (Q99496). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title