Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RNF2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
$243.50 - $524.00
Specifications
Antigen | RNF2 |
---|---|
Applications | ChIP Assay, Immunofluorescence, Immunohistochemistry, Immunoprecipitation, KnockDown |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Purified |
Description
RNF2 Polyclonal specifically detects RNF2 in Human, Mouse, Rat samples. It is validated for Western Blot, Chromatin Immunoprecipitation, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Knockout Validated.Specifications
RNF2 | |
Polyclonal | |
Purified | |
RUO | |
Human, Mouse, Rat | |
BAP-1, BAP1RING finger protein 1B, DINGRING finger protein BAP-1, EC 6.3.2, HIPI3RING1BE3 ubiquitin-protein ligase RING2, Huntingtin-interacting protein 2-interacting protein 3, Protein DinG, ring finger protein 2HIP2-interacting protein 3, RING1b, RING2EC 6.3.2.- | |
Recombinant fusion protein containing a sequence corresponding to amino acids 1-336 of human RNF2 (NP_009143.1). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
ChIP Assay, Immunofluorescence, Immunohistochemistry, Immunoprecipitation, KnockDown | |
Unconjugated | |
Rabbit | |
Apoptosis, Breast Cancer, Cancer, Signal Transduction, Transcription Factors and Regulators, Zinc Finger | |
PBS with 50% glycerol, pH7.3. | |
6045 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title