Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RNF20 Antibody, Alexa Fluor™ 350, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB1002243AF350
Description
RNF20 Polyclonal antibody specifically detects RNF20 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
RNF20 | |
Polyclonal | |
Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin | |
BRE1, BRE1 E3 ubiquitin ligase homolog, BRE1-A, BRE1AMGC129667, E3 ubiquitin-protein ligase BRE1A, EC 6.3.2, EC 6.3.2.-, FLJ11189, FLJ20382, hBRE1MGC129668, homolog of S. cerevisiae BRE1, KAIA2779, ring finger protein 20KIAA2779 | |
The immunogen recognized by this antibody maps to a region between residue 125 and 175 of human Ring Finger 20 using the numbering given in entry NP_062538.5 (GeneID 56254). | |
0.1 mL | |
Chromatin Research, Zinc Finger | |
56254 | |
Store at 4C in the dark. | |
IgG |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Alexa Fluor 350 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human, Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction