Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RNF20 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$487.50
Specifications
| Antigen | RNF20 |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
Description
RNF20 Polyclonal specifically detects RNF20 in Human samples. It is validated for Western Blot.Specifications
| RNF20 | |
| Polyclonal | |
| Rabbit | |
| Chromatin Research, Zinc Finger | |
| BRE1, BRE1 E3 ubiquitin ligase homolog, BRE1-A, BRE1AMGC129667, E3 ubiquitin-protein ligase BRE1A, EC 6.3.2, EC 6.3.2.-, FLJ11189, FLJ20382, hBRE1MGC129668, homolog of S. cerevisiae BRE1, KAIA2779, ring finger protein 20KIAA2779 | |
| RNF20 | |
| IgG |
| Western Blot | |
| Unconjugated | |
| RUO | |
| Q5VTR2 | |
| 56254 | |
| Synthetic peptides corresponding to RNF20(ring finger protein 20) The peptide sequence was selected from the middle region of RNF20. Peptide sequence KEREREREREKEKEREREKQKLKESEKERDSAKDKEKGKHDDGRKKEAEI. | |
| Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title