Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RNF20 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 13 publications
Supplier: Novus Biologicals NB1002242
Description
RNF20 Polyclonal specifically detects RNF20 in Human, Mouse samples. It is validated for Western Blot, Chromatin Immunoprecipitation, Immunoprecipitation.Specifications
RNF20 | |
Polyclonal | |
Unconjugated | |
Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide | |
BRE1, BRE1 E3 ubiquitin ligase homolog, BRE1-A, BRE1AMGC129667, E3 ubiquitin-protein ligase BRE1A, EC 6.3.2, EC 6.3.2.-, FLJ11189, FLJ20382, hBRE1MGC129668, homolog of S. cerevisiae BRE1, KAIA2779, ring finger protein 20KIAA2779 | |
Rabbit | |
Affinity Purified | |
RUO | |
56254 | |
Human, Mouse | |
IgG |
Western Blot, Immunoprecipitation, ChIP Assay | |
1.0 mg/mL | |
Western Blot 1:2000-1:10000, Immunoprecipitation 2-10 ug/mg lysate, Chromatin Immunoprecipitation (ChIP) 1:10-1:500 | |
Q5VTR2 | |
RNF20 | |
The immunogen recognized by this antibody maps to a region between residue 1 and 50 of human Ring Finger 20 using the numbering given in entry NP_062538.5 (GeneID 56254). | |
100 μL | |
Primary | |
Mouse reactivity reported in the scientific literature (PMID: 23760504). Based on 100% sequence identity, this antibody is predicted to react with Rat, Dog, Rabbit, Pig, Panda, Orangutan, Rhesus Monkey, Gorilla, Chimpanzee, Tasmanian devil, White-tufted-ear marmoset, Crab-eating macaque, European domestic ferret, Chinese hamster and Northern white-cheeked gibbon. | |
Store at 4C. Do not freeze. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction