Learn More
Invitrogen™ RNF20 Monoclonal Antibody (1C6)

Description
Peptide Sequence: RVESSRRAVS QIVTVYDKLQ EKVELLSRKL NSGDNLIVEE AVQELNSFLA QENMRLQELT DLLQEKHRTM SQEFSKLQSK VETAESRVSV LESMIDDLQW.
Specifications
Specifications
| Antigen | RNF20 |
| Applications | ELISA, Western Blot |
| Classification | Monoclonal |
| Clone | 1C6 |
| Concentration | 0.2-1.0 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with no preservative; pH 7.4 |
| Gene | RNF20 |
| Gene Accession No. | Q5VTR2 |
| Gene Alias | 4833430L21Rik; AW540162; BRE1; BRE1 E3 ubiquitin ligase homolog; BRE1A; BRE1-A; C79397; E3 ubiquitin-protein ligase BRE1A; FLJ11189; FLJ20382; hBRE1; HNF3B; homolog of S. cerevisiae BRE1; hypothetical protein; KIAA2779; Kiaa4116; MGC129667; MGC129668; mKIAA4116; RCJMB04_5a23; RING finger protein 20; ring finger protein 20, E3 ubiquitin protein ligase; RING-type E3 ubiquitin transferase BRE1A; Rnf20; TCF3B |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.