Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RNF20 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501988934
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-8934 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-8934 Supplier Bioss Supplier No. BS9243R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

RNF20 is an E3 ubiquitin ligase that regulates chromosome structure by monoubiquitinating histone H2B. RNF20 selectively suppresses the expression of some genes such as several protooncogenes and growth-related genes, including many genes that are induced by epidermal growth factor by interfering with chromatin recruitment of transcription elongation factor SII (TFIIS). In contrast, RNF20 also positively regulates the p53 tumor suppressor as well as numerous histone H2A and H2B genes. Together with the deubiquitinase USP44, RNF20 regulates stem cell differentiation by modulating H2B monoubiquitination.

Specifications

Antigen RNF20
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene RNF20
Gene Accession No. Q5VTR2
Gene Alias 4833430L21Rik; AW540162; BRE1; BRE1 E3 ubiquitin ligase homolog; BRE1A; BRE1-A; C79397; E3 ubiquitin-protein ligase BRE1A; FLJ11189; FLJ20382; hBRE1; HNF3B; homolog of S. cerevisiae BRE1; KIAA2779; Kiaa4116; MGC129667; MGC129668; mKIAA4116; RCJMB04_5a23; RING finger protein 20; ring finger protein 20, E3 ubiquitin protein ligase; RING-type E3 ubiquitin transferase BRE1A; RNF20; TCF3B
Gene Symbols RNF20
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human RNF20.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 56254
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.