Learn More
Invitrogen™ RNF20 Recombinant Rabbit Monoclonal Antibody (10H23L1)
Description
This antibody is predicted to react with Monkey, Cat, Pig and Bovine. Recombinant rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using recombinant rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire.
Specifications
Specifications
| Antigen | RNF20 |
| Applications | Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Clone | 10H23L1 |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.09% sodium azide; pH 7.2-7.4 |
| Gene | RNF20 |
| Gene Accession No. | Q5DTM8, Q5VTR2 |
| Gene Alias | 4833430L21Rik; AW540162; BRE1; BRE1 E3 ubiquitin ligase homolog; BRE1A; BRE1-A; C79397; E3 ubiquitin-protein ligase BRE1A; FLJ11189; FLJ20382; hBRE1; HNF3B; homolog of S. cerevisiae BRE1; hypothetical protein; KIAA2779; Kiaa4116; MGC129667; MGC129668; mKIAA4116; RCJMB04_5a23; RING finger protein 20; ring finger protein 20, E3 ubiquitin protein ligase; RING-type E3 ubiquitin transferase BRE1A; Rnf20; TCF3B |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.