Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RNF211 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501988599
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-8599 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-8599 Supplier Bioss Supplier No. BS8495R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

RFPL4B gene ontology (GO) annotation includes metal ion binding.

Specifications

Antigen RNF211
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene RFPL4B
Gene Accession No. Q6ZWI9
Gene Alias ret finger protein like 4B; Ret finger protein-like 4B; RFPL4B; RING finger protein 211; RNF211
Gene Symbols RFPL4B
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human RFPL4B/RNF211.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 442247
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.