Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RNF212 Monoclonal Antibody (5H3)
GREENER_CHOICE

Catalog No. PIMA523246
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA523246 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA523246 Supplier Invitrogen™ Supplier No. MA523246
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Peptide Sequence: IKSSVSTKPH GCLLPPHSSA PDRLESMEVD LSPSPIRKSE IAAGPARISM ISPPQDGRMA PCARRVCHFQ RFTMFLHRRL SSLAAPPSVQ FWKARGTHQL.

This gene encodes a RING finger protein that may function as a ubiquitin ligase. The encoded protein may be involved in meiotic recombination. This gene is located within a linkage disequilibrium block and polymorphisms in this gene may influence recombination rates. Alternate splicing results in multiple transcript variants.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RNF212
Applications ELISA, Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 5H3
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative; pH 7.4
Gene RNF212
Gene Accession No. Q495C1
Gene Alias EG330157; F830003B07; Probable E3 SUMO-protein ligase RNF212; Probable E3 SUMO-protein transferase RNF212; RING finger protein 212; RNF212; ZHP3
Gene Symbols RNF212
Host Species Mouse
Immunogen RNF212 (NP_919420, 133 a.a. ~ 232 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 285498
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.