Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RNF212 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA563448
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA563448 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA563448 Supplier Invitrogen™ Supplier No. PA563448
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: KLLEFIKHVC YHRHQSHRPC APGWFCQVLQ RPGAVSGEKT QQTRPAPPAT CLLCLSCLSG FRHGPWRSQA LPSDLVAPLF VSYTVEVSIT NAGWS Highest antigen sequence identity to the following orthologs: Mouse - 27%, Rat - 24%.

This gene encodes a RING finger protein that may function as a ubiquitin ligase. The encoded protein may be involved in meiotic recombination. This gene is located within a linkage disequilibrium block and polymorphisms in this gene may influence recombination rates. Alternate splicing results in multiple transcript variants.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RNF212
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene RNF212
Gene Accession No. Q495C1
Gene Alias EG330157; F830003B07; Probable E3 SUMO-protein ligase RNF212; Probable E3 SUMO-protein transferase RNF212; RING finger protein 212; RNF212; ZHP3
Gene Symbols RNF212
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human RNF212. Recombinant protein control fragment (Product #RP-103805).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 285498
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.