Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ RNF212 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5112697
Description
RNF212 Polyclonal Antibody for Western Blot, IHC (P), ELISA
This gene encodes a RING finger protein that may function as a ubiquitin ligase. The encoded protein may be involved in meiotic recombination. This gene is located within a linkage disequilibrium block and polymorphisms in this gene may influence recombination rates. Alternate splicing results in multiple transcript variants.
Specifications
| RNF212 | |
| Polyclonal | |
| Unconjugated | |
| RNF212 | |
| EG330157; F830003B07; Probable E3 SUMO-protein ligase RNF212; Probable E3 SUMO-protein transferase RNF212; RING finger protein 212; RNF212; ZHP3 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 285498 | |
| -20°C or -80°C if preferred | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot | |
| 1.43 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q495C1 | |
| RNF212 | |
| Recombinant Human Probable E3 SUMO-protein ligase RNF212 protein (1-297AA). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction