Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RNF213 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA518650
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA518650 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA518650 Supplier Invitrogen™ Supplier No. PA518650
Add to Cart
Add to Cart

Goat Polyclonal Antibody

This antibody is tested in Peptide ELISA: antibody detection limit dilution 4,000.

RNF213 is an E3 ubiquitin-protein ligase involved in angiogenesis. Involved in the non-canonical Wnt signaling pathway in vascular development: acts by mediating ubiquitination and degradation of FLNA and NFATC2 downstream of RSPO3, leading to inhibit the non-canonical Wnt signaling pathway and promoting vessel regression. Also has ATPase activity.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RNF213
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3
Gene RNF213
Gene Accession No. Q63HN8
Gene Alias 6030403J01; ALK lymphoma oligomerization partner on chromosome 17; ALO17; C17orf27; D11Ertd759e; E3 ubiquitin-protein ligase RNF213; KIAA1554; KIAA1618; mKIAA1554; MYMY2; Mysterin; MYSTR; NET57; protein ALO17; RGD1308168; RING finger protein 213; RING-type E3 ubiquitin transferase RNF213; RNF213
Gene Symbols RNF213
Host Species Goat
Immunogen Synthetic peptide sequence (KTAAVLKWNRE) corresponding to the C-terminus amino acids of RNF213 (aa 5195-5206).
Purification Method Ammonium sulfate precipitation
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57674
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.