Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RNF213 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA555418
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA555418 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA555418 Supplier Invitrogen™ Supplier No. PA555418
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: KDPVCLPCDH VHCLRCLRAW FASEQMICPY CLTALPDEFS PAVSQAHREA IEKHARFRQM CNSFFVDLVS TICFKDNAPP EKEVIESLLS LLFVQKGRLR DAAQRHCEHT KSLSPFNDVV DKTPVIRSVI LKLLLKYS Highest antigen sequence identity to the following orthologs: Mouse - 78%, Rat - 79%.

RNF213 is an E3 ubiquitin-protein ligase involved in angiogenesis. Involved in the non-canonical Wnt signaling pathway in vascular development: acts by mediating ubiquitination and degradation of FLNA and NFATC2 downstream of RSPO3, leading to inhibit the non-canonical Wnt signaling pathway and promoting vessel regression. Also has ATPase activity.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RNF213
Applications Western Blot
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene RNF213
Gene Accession No. Q63HN8
Gene Alias 6030403J01; ALK lymphoma oligomerization partner on chromosome 17; ALO17; C17orf27; D11Ertd759e; E3 ubiquitin-protein ligase RNF213; KIAA1554; KIAA1618; mKIAA1554; MYMY2; Mysterin; MYSTR; NET57; protein ALO17; RGD1308168; RING finger protein 213; RING-type E3 ubiquitin transferase RNF213; RNF213
Gene Symbols RNF213
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human RNF213. Recombinant protein control fragment (Product #RP-89516).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57674
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.