Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RNF213 Rabbit anti-Human, Unconjugated, Polyclonal, MilliporeSigma™

Catalog No. ABN1474100 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ABN1474-100 100 μg
ABN1474-25UG 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ABN1474-100 Supplier MilliporeSigma Supplier No. ABN1474100UG
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Anti-RNF213, Cat. No. ABN1474, is a highly specific rabbit polyclonal antibody that targets E3 ubiquitin-protein ligase RNF213 and has been tested for use in Immunohistochemistry (Paraffin) and Western Blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RNF213
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Formulation Purified rabbit polyclonal antibody in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.
Gene Alias E3 ubiquitin-protein ligase RNF213;EC:2.3.2.27;ALK lymphoma oligomerization partner on chromosome 17;Mysterin;RING finger protein 213;RING-type E3 ubiquitin transferase RNF213
Gene Symbols RNF213;ALO17;C17orf27;KIAA1554;KIAA1618;MYSTR
Host Species Rabbit
Immunogen KLH-conjugated linear peptide corresponding to 15 amino acids from the C-terminal half of human E3 ubiquitin-protein ligase RNF213. Immunogen sequence is conserved in isoforms 1, 2, and 4.
Molecular Weight of Antigen ∽590 kDa observed; 591.41 kDa calculated. Uncharacterized bands may be observed in some lysate(s).
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) NP_001243000
Test Specificity This rabbit polyclonal antibody detects E3 ubiquitin-protein ligase RNF213 in human cells. It targets an epitope within 15 amino acids from the C-terminal half.
Target Species Human
Content And Storage Stable for 1 year at 2-8°C from date of receipt.
Product Type Antibodies
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.