Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ RNF214 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA550874
Description
The antibody detects endogenous levels of total RNF214 protein.
RNF214 (RING finger protein 214) is a 703 amino acid protein that contains one RING-type zinc finger and is encoded by a gene that maps to human chromosome 11q23.3. Chromosome 11 houses over 1,400 genes and comprises nearly 4% of the human genome. Jervell and Lange-Nielsen syndrome, Jacobsen syndrome, Niemann-Pick disease, hereditary angioedema and Smith-Lemli-Opitz syndrome are associated with defects in genes that maps to chromosome 11.
Specifications
RNF214 | |
Polyclonal | |
Unconjugated | |
RNF214 | |
D130054N24Rik; ring finger protein 214; Rnf214 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
257160 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1.2 mg/mL | |
PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
Q8ND24 | |
RNF214 | |
Fusion protein corresponding to residues near the C terminal of human ring finger protein 214. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction