Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RNF214 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA558739
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA558739 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA558739 Supplier Invitrogen™ Supplier No. PA558739
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: VTRSLKAGCH TKQLASRNCS EEKSPQTSIL KEGNRDTSLD FRPVVSPANG VEGVRVDQDD DQDSSSLKLS QNIAVQTDFK TADSEVNTDQ DIEKNLD Highest antigen sequence identity to the following orthologs: Mouse - 88%, Rat - 93%.

RNF214 (RING finger protein 214) is a 703 amino acid protein that contains one RING-type zinc finger and is encoded by a gene that maps to human chromosome 11q23.3. Chromosome 11 houses over 1,400 genes and comprises nearly 4% of the human genome. Jervell and Lange-Nielsen syndrome, Jacobsen syndrome, Niemann-Pick disease, hereditary angioedema and Smith-Lemli-Opitz syndrome are associated with defects in genes that maps to chromosome 11.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RNF214
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene RNF214
Gene Accession No. Q8ND24
Gene Alias D130054N24Rik; ring finger protein 214; Rnf214
Gene Symbols RNF214
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human RNF214. Recombinant protein control fragment (Product #RP-97451).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 257160
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.