Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RNF215 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$382.00
Specifications
Antigen | RNF215 |
---|---|
Dilution | Immunohistochemistry 1:20 - 1:50, Immunohistochemistry-Paraffin 1:20 - 1:50 |
Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
RNF215 Polyclonal antibody specifically detects RNF215 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
RNF215 | |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Rabbit | |
Zinc Finger | |
PBS (pH 7.2) and 40% Glycerol | |
200312 | |
IgG | |
Protein A purified |
Immunohistochemistry 1:20 - 1:50, Immunohistochemistry-Paraffin 1:20 - 1:50 | |
Polyclonal | |
Purified | |
RUO | |
Human | |
ring finger protein 215 | |
This antibody was developed against a recombinant protein corresponding to amino acids: LPLLVQFPAMGNSYGAMTCLTRWLRRGQKGLDDVSRAANCTSSCQLCKWICPSALGVWRGWENPGHLISNPVPRWVPSTPMPWGPGQWAAGGCVLGTSPMVWFGDPFQEPRRGTHRSRARISM | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title