Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RNF215 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA566019
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA566019 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA566019 Supplier Invitrogen™ Supplier No. PA566019
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LDISQLLLRPV IVLHYSSNVT KLLDALLQRT QATAEITSGE SLSANIEWKL TLWTTCGLSK DGYGGWQDLV CLGGSRAQEQ KP Highest antigen sequence identity to the following orthologs - mouse 94%, rat 95%.

RNF215 gene ontology annotations related to this gene include ubiquitin-dependent protein catabolic process.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RNF215
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene RNF215
Gene Accession No. Q9Y6U7
Gene Alias 0610009J22Rik; AW557025; C77903; RGD1310738; ring finger protein 215; Rnf215
Gene Symbols RNF215
Host Species Rabbit
Immunogen Recombinant Human RNF215. Recombinant protein control fragment (Product #RP-106703).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 200312
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.