Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RNF215 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA548303
Description

Specifications
RNF215 | |
Polyclonal | |
PBS with 0.09% sodium azide | |
Q9Y6U7, Q5SPX3 | |
RNF215 | |
KLH conjugated synthetic peptide between 266-295 amino acids from the C-terminal region of human RNF215. | |
400 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
RNF215 | |
RING finger protein 215, RNF215 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
200312, 71673 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction