Learn More
Invitrogen™ RNF216 Polyclonal Antibody

Description
Suggested positive control: human spleen protein, antigen standard for RNF216 (transient overexpression lysate), human lung protein, human ovary protein, human pancreas protein, human skeletal muscle protein, human heart protein, human kidney protein, human liver protein, human brain protein, human brain tissue, heart tissue, kidney tissue, liver tissue, lung tissue, ovary tissue, pancreas tissue, small intestine tissue, skeletal muscle tissue, spleen tissue, human small intestine protein.
Specifications
Specifications
| Antigen | RNF216 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.2% gelatin and 0.05% sodium azide |
| Gene | RNF216 |
| Gene Accession No. | Q9NWF9 |
| Gene Alias | 2810055G22Rik; AI647468; AU019462; C86502; CAHH; E3 ubiquitin ligase; E3 ubiquitin-protein ligase RNF216; F830018F18Rik; RGD1565135; RING finger protein 216; RING-type E3 ubiquitin transferase RNF216; RNF216; Triad domain-containing protein 3; TRIAD3; triad3a; U7I1; UBCE7IP1; UbcM4-interacting protein 83; ubiquitin-conjugating enzyme 7-interacting protein 1; Uip83; Zin; zinc finger protein inhibiting NF-kappa-B |
| Gene Symbols | RNF216 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.