Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RNF217 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA542360
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA542360 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA542360 Supplier Invitrogen™ Supplier No. PA542360
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: GLALGAIAVV IVEEIKTYWN LISGRTRNQT QHLAPQPVLL SDMLYCLKQV Sequence homology: Human: 100%; Yeast: 91%.

RNF217 is an E3 ubiquitin-protein ligase which accepts ubiquitin from E2 ubiquitin-conjugating enzymes in the form of a thioester and then directly transfers the ubiquitin to targeted substrates.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RNF217
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene RNF217
Gene Accession No. Q8TC41
Gene Alias C6orf172; dJ84N20.1; IBR domain containing 1; IBR domain-containing protein 1; IBRDC1; opposite STL; OSTL; Probable E3 ubiquitin-protein ligase RNF217; ring finger protein 217; RNF217
Gene Symbols RNF217
Host Species Rabbit
Immunogen Synthetic peptide directed towards the middle region of human RNF217.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 154214
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.