Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RNF219 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP140358
Description
RNF219 Polyclonal specifically detects RNF219 in Human samples. It is validated for Western Blot, Immunoprecipitation.Specifications
RNF219 | |
Polyclonal | |
Unconjugated | |
TBS and 0.1% BSA with 0.09% Sodium Azide | |
RNF219 | |
The immunogen for this product maps to a region between residue 676 and 726 of human ring finger protein 219 using the numbering given in entry NP_078822.3 (GeneID 79596). | |
0.1 mL | |
Primary | |
Based on 100% sequence identity, this antibody is predicted to react with Tasmanian devil and Gray short-tailed opossum. | |
Store at 4C. Do not freeze. |
Western Blot, Immunoprecipitation | |
0.2 mg/mL | |
Western Blot 1:2000-1:10000, Immunoprecipitation 5-15 ug/mg lysate | |
C13orf7, chromosome 13 open reading frame 7, DKFZp686A01276, DKFZp686N15250, DKFZp686O03173, FLJ13449, FLJ25774, ring finger protein 219 | |
Rabbit | |
Affinity Purified | |
RUO | |
79596 | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction