Learn More
Invitrogen™ RNF220 Monoclonal Antibody (5E5)
Mouse Monoclonal Antibody
Supplier:  Invitrogen™ MA524470
Catalog No. PIMA524470
Only null left
Add to Cart
Description
Description
Peptide Sequence: QEAMQKTCKN SDIEKITEDS AVTTFEALKA RVRELERQLS RGDRYKCLIC MDSYSMPLTS IQCWHVHCEE CWLRTLGAKK LCPQCNTITA PGDLRRIYL.
E3 ubiquitin-protein ligase that promotes the ubiquitination and proteasomal degradation of SIN3B. Independently of its E3 ligase activity, acts as a CTNNB1 stabilizer through USP7-mediated deubiquitination of CTNNB1 promoting Wnt signaling (PubMed:25266658).
TRUSTED_SUSTAINABILITY
Specifications
Specifications
RNF220
Monoclonal
Unconjugated
RNF220
4732477A13; 4931406I20Rik; 5730503K05Rik; AV302541; C1orf164; E3 ubiquitin-protein ligase RNF220; ring finger protein 220; RING-type E3 ubiquitin transferase Rnf220; RNF220
Mouse
Protein A
RUO
55182
-20°C, Avoid Freeze/Thaw Cycles
Liquid
ELISA, Immunocytochemistry
5E5
PBS with no preservative; pH 7.4
Q5VTB9
RNF220
FLJ10597 (NP_060620, 468 a.a. ~ 566 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
100 μg
Primary
Human
Antibody
IgG1 κ
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.