Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ RNF220 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA565014
Description
Immunogen sequence: RGSGFIMCSG KENPDSDADL DVDGDDTLEY GKPQYTEADV IPCTGEEPGE AKEREALRGA VLNGGPPSTR ITPEFSKWAS.
E3 ubiquitin-protein ligase that promotes the ubiquitination and proteasomal degradation of SIN3B. Independently of its E3 ligase activity, acts as a CTNNB1 stabilizer through USP7-mediated deubiquitination of CTNNB1 promoting Wnt signaling (PubMed:25266658).
Specifications
RNF220 | |
Polyclonal | |
Unconjugated | |
RNF220 | |
4732477A13; 4931406I20Rik; 5730503K05Rik; AV302541; C1orf164; E3 ubiquitin-protein ligase RNF220; ring finger protein 220; RING-type E3 ubiquitin transferase Rnf220; RNF220 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
55182 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot, Immunocytochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q5VTB9 | |
RNF220 | |
Recombinant Human RNF220. Recombinant protein control fragment (Product #RP-104582). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction