Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RNF220 Polyclonal Antibody
RNF220 Polyclonal Antibody
Supplier: Thermo Scientific PA5110308
Description
RNF220 Polyclonal Antibody for Western Blot

Specifications
RNF220 | |
Polyclonal | |
Unconjugated | |
RNF220 | |
4732477A13; 4931406I20Rik; 5730503K05Rik; AV302541; C1orf164; E3 ubiquitin-protein ligase RNF220; ring finger protein 220; RING-type E3 ubiquitin transferase Rnf220; RNF220 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
500532, 55182, 66743 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot | |
1.86 mg/mL | |
PBS with 50% glycerol and 0.05% ProClin 300; pH 7.3 | |
Q5VTB9, Q6PDX6 | |
RNF220 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 267-566 of human RNF220 (NP_060620.2). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction