Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RNF220 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA555660
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA555660 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA555660 Supplier Invitrogen™ Supplier No. PA555660
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: QPRPFGVPVS VDKDVHIPFT NGSYTFASMY HRQGGVPGTF ANRDFPPSLL HLHPQFAPPN LDCTPISMLN HSGVGAFRPF ASTEDRESYQ SAFTPAKRLK NC Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 27%.

E3 ubiquitin-protein ligase that promotes the ubiquitination and proteasomal degradation of SIN3B. Independently of its E3 ligase activity, acts as a CTNNB1 stabilizer through USP7-mediated deubiquitination of CTNNB1 promoting Wnt signaling (PubMed:25266658).
TRUSTED_SUSTAINABILITY

Specifications

Antigen RNF220
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene RNF220
Gene Accession No. Q5VTB9, Q6PDX6
Gene Alias 4732477A13; 4931406I20Rik; 5730503K05Rik; AV302541; C1orf164; E3 ubiquitin-protein ligase RNF220; ring finger protein 220; RING-type E3 ubiquitin transferase Rnf220; RNF220
Gene Symbols RNF220
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human RNF220. Recombinant protein control fragment (Product #RP-94522).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55182, 66743
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.