Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RNF224 Polyclonal Antibody
GREENER_CHOICE

Catalog No. FEPA5145199
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEPA5145199 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEPA5145199 Supplier Invitrogen™ Supplier No. PA5145199
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PARLEPLPLT SLKGSAITRQ PAGLCPALGP QPHFPQPRYC CWGCGSLCCP PLGSPEV Antigen sequence identity: Mouse 60%; Rat 62%.

RNF224 is a protein coding gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RNF224
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene RNF224
Gene Accession No. P0DH78
Gene Alias RING finger protein 224; RNF224
Gene Symbols RNF224
Host Species Rabbit
Immunogen Recombinant Protein Epitope Signature Tag (PrEST) antigen sequence. Antigen range: 100-156. Recombinant protein control fragment (Product #RP-110108).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 643596
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Format Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.