Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RNF23 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA540967
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA540967 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA540967 Supplier Invitrogen™ Supplier No. PA540967
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: HTVVPLDDAT QEYKEKLQKC LEPLEQKLQE ITRCKSSEEK KPGELKRLVE Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 93%; Rabbit: 100%; Rat: 100%.

The protein encoded by this gene is a member of the tripartite motif (TRIM) family. The TRIM motif includes three zinc-binding domains, a RING, a B-box type 1 and a B-box type 2, and a coiled-coil region. The function of this protein has not been identified. This gene lies within the major histocompatibility complex class I region on chromosome 6. Alternate splicing results in two transcript variants encoding different isoforms.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RNF23
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene TRIM39
Gene Accession No. Q9HCM9
Gene Alias 1100001D15Rik; E130103K13Rik; E3 ubiquitin-protein ligase TRIM39; mKIAA4179; RBCC-B30.2; ring finger protein 23; RING-B box-coiled coil-B30.2; RING-B box-coiled-coil-B30.2; RING-type E3 ubiquitin transferase TRIM39; Rnf23; Testis-abundant finger protein; tfp; TRIM39; TRIM39B; tripartite motif containing 39; tripartite motif protein 39; tripartite motif-containing 39; tripartite motif-containing protein 39
Gene Symbols TRIM39
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal of human TRIM39 (aa 138-187).
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 56658
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.