Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RNF23 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA530771
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA530771 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA530771 Supplier Invitrogen™ Supplier No. PA530771
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: A431, mouse testis, rat testis. Predicted reactivity: Mouse (97%), Rat (96%), Pig (99%), Chimpanzee (87%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

The protein encoded by this gene is a member of the tripartite motif (TRIM) family. The TRIM motif includes three zinc-binding domains, a RING, a B-box type 1 and a B-box type 2, and a coiled-coil region. The function of this protein has not been identified. This gene lies within the major histocompatibility complex class I region on chromosome 6. Alternate splicing results in two transcript variants encoding different isoforms.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RNF23
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7
Gene TRIM39
Gene Accession No. Q6MFZ5, Q9ESN2, Q9HCM9
Gene Alias 1100001D15Rik; E130103K13Rik; E3 ubiquitin-protein ligase TRIM39; mKIAA4179; RBCC-B30.2; ring finger protein 23; RING-B box-coiled coil-B30.2; RING-B box-coiled-coil-B30.2; RING-type E3 ubiquitin transferase TRIM39; Rnf23; Testis-abundant finger protein; tfp; TRIM39; TRIM39B; tripartite motif containing 39; tripartite motif protein 39; tripartite motif-containing 39; tripartite motif-containing protein 39
Gene Symbols TRIM39
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human RNF23. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 309591, 56658, 79263
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.