Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RNF24 Monoclonal Antibody (4C6)
GREENER_CHOICE

Catalog No. PIMA521310
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA521310 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA521310 Supplier Invitrogen™ Supplier No. MA521310
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Peptide Sequence: LRHQAHKEFY AYKQVILKEK VKELNLHELC AVCLEDFKPR DELGICPCKH AFHRKCLIKW LEVRKVCPLC NMPVLQLAQL HSKQDRGPPQ GPLPGAENIV.

The protein encoded by this gene contains similarity to the Drosophila goliath protein and thus may function as a transcription factor. Multiple alternatively spliced transcript variants that encode different isoforms have been found for this gene. [provided by RefSeq, Aug 2008].
TRUSTED_SUSTAINABILITY

Specifications

Antigen RNF24
Applications ELISA, Immunocytochemistry
Classification Monoclonal
Clone 4C6
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative; pH 7.4
Gene RNF24
Gene Accession No. Q9Y225
Gene Alias 2810473M14Rik; 4930505A13Rik; AI317164; C86507; D2Ertd504e; G1L; RING finger protein 24; Rnf24
Gene Symbols RNF24
Host Species Mouse
Immunogen RNF24 (NP_009150, 49 a.a. ~ 148 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity Chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11237
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.