Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RNF24 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA552872
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA552872 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA552872 Supplier Invitrogen™ Supplier No. PA552872
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: IRLRHQAHKE FYAYKQVILK EKVKELNLHE LCAVCLEDFK PRDELGICPC KHAFHRKCLI KWLEVRKVCP LCNMPVLQLA QLHSKQDRG Highest antigen sequence identity to the following orthologs: Mouse - 99%, Rat - 54%.

The protein encoded by this gene contains similarity to the Drosophila goliath protein and thus may function as a transcription factor. Multiple alternatively spliced transcript variants that encode different isoforms have been found for this gene. [provided by RefSeq, Aug 2008].
TRUSTED_SUSTAINABILITY

Specifications

Antigen RNF24
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene RNF24
Gene Accession No. Q9Y225
Gene Alias 2810473M14Rik; 4930505A13Rik; AI317164; C86507; D2Ertd504e; G1L; RING finger protein 24; Rnf24
Gene Symbols RNF24
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human RNF24. Recombinant protein control fragment (Product #RP-90034).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11237
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.