Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ RNF25 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA528080
Description
Recommended positive controls: H1299. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
The protein encoded by this gene contains a RING finger motif. The mouse counterpart of this protein has been shown to interact with Rela, the p65 subunit of NF-kappaB (NFKB), and modulate NFKB-mediated transcription activity. The mouse protein also binds ubiquitin-conjugating enzymes (E2s) and is a substrate for E2-dependent ubiquitination.
Specifications
RNF25 | |
Polyclonal | |
Unconjugated | |
RNF25 | |
0610009H16Rik; AO7; E3 ubiquitin-protein ligase RNF25; ring finger protein 25; RING finger protein AO7; RING-type E3 ubiquitin transferase RNF25; RNF25 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
64320 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
1 mg/mL | |
PBS with 40% glycerol and 0.01% thimerosal; pH 7 | |
Q96BH1 | |
RNF25 | |
Carrier-protein conjugated synthetic peptide encompassing a sequence within the C-terminus region of human RNF25. The exact sequence is proprietary. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction