Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RNF25 Polyclonal Antibody

RNF25 Polyclonal Antibody
Supplier: Thermo Scientific PA557650
Description
RNF25 Polyclonal Antibody for Western Blot, ICC/IF, IHC
Specifications
| RNF25 | |
| Polyclonal | |
| Unconjugated | |
| RNF25 | |
| 0610009H16Rik; AO7; E3 ubiquitin-protein ligase RNF25; ring finger protein 25; RING finger protein AO7; RING-type E3 ubiquitin transferase RNF25; RNF25 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 64320 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.5 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q96BH1 | |
| RNF25 | |
| Recombinant protein corresponding to Human RNF25. Recombinant protein control fragment (Product #RP-96183). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction