Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RNF25 Polyclonal Antibody, Bethyl Laboratories

Catalog No. 501566207
Encompass_Preferred
Change view
Click to view available options
Quantity:
20 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-156-6207 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-156-6207 Supplier Bethyl Laboratories, Inc Supplier No. A303844AT
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The recommended shelf life for this product is 1 year from date of receipt.

The protein encoded by this gene contains a RING finger motif. The mouse counterpart of this protein has been shown to interact with Rela, the p65 subunit of NF-kappaB (NFKB), and modulate NFKB-mediated transcription activity. The mouse protein also binds ubiquitin-conjugating enzymes (E2s) and is a substrate for E2-dependent ubiquitination.

Specifications

Antigen RNF25
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation TBS with 0.1% BSA and 0.09% sodium azide
Gene RNF25
Gene Accession No. Q96BH1
Gene Alias AO7, E3 ubiquitin-protein ligase RNF25, ring finger protein AO7
Gene Symbols RNF25
Host Species Rabbit
Immunogen Between 409 and 459
Purification Method Antigen affinity chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 64320
Target Species Human
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.