Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RNF26 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA572894
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA572894 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA572894 Supplier Invitrogen™ Supplier No. PA572894
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

RNF26 Polyclonal Antibody for Western Blot, ELISA

The protein encoded by this intronless gene contains a C3HC5 type of RING finger, a motif known to be involved in protein-DNA and protein-protein interactions. The expression of this gene was found to be upregulated in cancer cell lines derived from different types of cancer.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RNF26
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene RNF26
Gene Accession No. Q9BY78
Gene Alias 8030450I18Rik; E3 ubiquitin-protein ligase RNF26; RING finger protein 26; ring finger protein with leucine zipper; Rnf26
Gene Symbols RNF26
Host Species Rabbit
Immunogen RNF26 antibody for a peptide corresponding to 14 amino acids near the amino terminus of human RNF26.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 300659, 79102
Target Species Human, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.