Learn More
Invitrogen™ RNF3 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5113596
Description
Immunogen sequence: FYHKLGMEVP GDIKGETCSA KQHLDSHRNG ETKADDSSNK EAAEEKPEED For short term use, store at 2-8°C up to 1 week. For long term storage, store at -20°C in small aliquots to prevent freeze-thaw cycles. Predicted homology: Cow: 93%; Dog: 93%; Guinea Pig: 86%; Horse: 100%; Human: 100%; Mouse: 86%; Pig: 100%; Rabbit: 86%; Rat: 86%.
Phosphorylase kinase is a hexadecameric enzyme that is comprised of four copies of four subunits that are encoded by four separate genes: PHKA, PHKB, PHKG, and PH kDa. This serine/threonine specific kinase converts glycogen phosphorylase b to glycogen phosphorylase a, resulting in the release of glucose-1-phophate from glycogen. PHKB (Phosphorylase b kinase regulatory subunit beta) is a 1093 amino acid subunit of phosphorylase kinase that, along with PHKA, has regulatory functions controlled by phosphorylation. Defects in the gene encoding PHKB are the cause of glycogen storage disease type 9B, which is also known as phosphorylase kinase deficiency of liver and muscle. This disease is characterized by a mild phenotype of hepatomegaly with only slightly elevated transaminase and plasma lipids, no clinical muscle involvement, and generally is correlated with a gradual improvement with increasing age. There are four isoforms of PHKB that are produced as a result of alternative splicing events.
Specifications
RNF3 | |
Polyclonal | |
Unconjugated | |
PCGF3 | |
2310035N15Rik; AI662857; D630042K08Rik; D6S113E; DADB-17J1.3; DKFZp686D20235; DKFZp686N0336; DONG1; E430039C14; FLJ31942; FLJ36550; FLJ43813; FSH; FSRG1; KIAA9001; MGC129075 protein; MGC129615; MGC40413; NAT; PCGF3; polycomb group ring finger 3; polycomb group RING finger protein 3; ring finger protein 3; RING finger protein 3A; RING3; RNF3; RNF3A | |
Rabbit | |
Affinity Chromatography | |
RUO | |
10336 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q3KNV8 | |
PCGF3 | |
Synthetic peptide directed towards the middle region of human PCGF3. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.