Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ RNF31 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5110309
Description
Immunogen sequence: AAGACPEEIF SALQYSGTEV PLQWLRSELP YVLEMVAELA GQQDPGLGAF SCQEARRAWL DRHGNLDEAV EECVRTRRRK VQELQSLGFG PEEGSLQALF QHGGDVSRAL TELQRQRLEP FRQRLWDSGP EPTPSWDGPD KQSLVRRLLA VYALPSWGRA ELALSLLQET PRNYELGDVV EAVRHSQDRA FLRRLLAQEC A.
Lyn proto oncogene (Lcl/Yes related novel protein tyrosine kinase) is encoded by the Lyn gene located on chromosome 8 in humans and belongs to the Src family kinase. It is a non-receptor tyrosine kinase that acts as a mediator in transducing cellular signaling. The most well studied expression of Lyn is in hematopoietic cell types, both lymphoid and myeloid cells with exclusion in T lymphocytes. It has also been observed that Lyn plays diverse roles such as in regulation of B cell receptor signaling, mast cell signaling and Epithelial - Mesenchymal Transition (EMT).
Specifications
RNF31 | |
Polyclonal | |
Unconjugated | |
RNF31 | |
AL033293; BC031509; E3 ubiquitin-protein ligase RNF31; Flj10111; FLJ23501; HOIL-1-interacting protein; HOIP; mFLJ00217; MGC19975; Paul; putative Ariadne-like ubiquitin ligase; RING finger protein 31; RING-type E3 ubiquitin transferase RNF31; Rnf31; ZIBRA; Zinc in-between-RING-finger ubiquitin-associated domain protein | |
Rabbit | |
Affinity chromatography | |
RUO | |
268749, 55072 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
1.54 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q924T7, Q96EP0 | |
RNF31 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 500-700 of human RNF31 (NP_060469.4). | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction