Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RNF32 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA542342
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA542342 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA542342 Supplier Invitrogen™ Supplier No. PA542342
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: ACLQAFEKFT NKKTCPLCRK NQYQTRVIHD GARLFRIKCV TRIQAYWRGC Sequence homology: Cow: 93%; Dog: 93%; Guinea Pig: 93%; Horse: 100%; Human: 100%; Mouse: 100%; Pig: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 79%.

RNF32 contains two RING ring finger motifs. RING finger motifs are present in a variety of functionally distinct proteins and are known to be involved in protein-DNA or protein-protein interactions. Its gene was found to be expressed during spermatogenesis, most likely in spermatocytes and/or in spermatids.The protein encoded by this gene contains two RING ring finger motifs. RING finger motifs are present in a variety of functionally distinct proteins and are known to be involved in protein-DNA or protein-protein interactions. This gene was found to be expressed during spermatogenesis, most likely in spermatocytes and/or in spermatids. Several alternatively spliced transcript variants exist, but their full length natures are not clear.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RNF32
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene RNF32
Gene Accession No. Q9H0A6
Gene Alias 1700009J01Rik; 2700025B22Rik; 4930542N22Rik; FKSG33; HSD15; Limb region protein 2; Lmbr2; RING finger protein 32; RNF32; tcag7.575; zgc:153042
Gene Symbols RNF32
Host Species Rabbit
Immunogen Synthetic peptide directed towards the middle region of human RNF32.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 140545
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.