Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ RNF34 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA567332
Description
Immunogen sequence: NIVCKACGLSF SVFRKKHVCC DCKKDFCSVC SVLQENLRRC STCHLLQETA FQRPQLMRLK VKDL Highest antigen sequence identity to the following orthologs - mouse 97%, rat 97%.
The protein encoded by this gene contains a RINF finger, a motif known to be involved in protein-protein and protein-DNA interactions. This protein interacts with DNAJA3/hTid-1, which is a DnaJ protein reported to function as a modulator of apoptosis. Overexpression of this gene in Hela cells was shown to confer the resistance to TNF-alpha induced apoptosis, suggesting an anti-apoptotic function of this protein. This protein can be cleaved by caspase-3 during the induction of apoptosis. Alternatively spliced transcript variants encoding distinct isoforms have been reported.
Specifications
RNF34 | |
Polyclonal | |
Unconjugated | |
RNF34 | |
AW061037; AW536122; BC004042; C88279; CARP1; CARP-1; Caspase regulator CARP1; caspases-8 and -10-associated RING finger protein 1; E3 ubiquitin-protein ligase RNF34; FLJ21786; FYVE-RING finger protein MOMO; hRFI; Human RING finger homologous to inhibitor of apoptosis protein; Momo; phafin 1; phafin-1; RFI; RIF; RIFF; ring finger protein 34; ring finger protein 34, E3 ubiquitin protein ligase; RING finger protein MOMO; RING finger protein RIFF; RING-type E3 ubiquitin transferase RNF34; RNF34 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
80196 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot, Immunocytochemistry | |
0.16 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q969K3 | |
RNF34 | |
Recombinant Human RNF34. Recombinant protein control fragment (Product #RP-105587). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction