Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RNF34 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA597100
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA597100 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA597100 Supplier Invitrogen™ Supplier No. PA597100
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MKAGATSMWA SCCGLLNEVM GTGAVRGQQS AFAGATGPFR FTPNPEFSTY PPAATEGPNI VCKACGLSFS VFRKKHVCCD CKKDFCSVCS VLQENLRRCS TCHLLQETAF QRPQLMRLKV KDLRQYLILR NIPIDTCREK EDLVDLVLCH HGLGSEDDMD TSSLNSSRSQ TSSFFTRSFF SNYTAPSATM SSFQGELMDG DQTSRSGVPA QVQSEITSAN; Positive Samples: U-251MG, LO2, HeLa, BT-474; Cellular Location: Cell membrane, Cytoplasm, Endomembrane system, Nucleus, Nucleus speckle, Peripheral membrane protein, cytosol.

The protein encoded by this gene contains a RINF finger, a motif known to be involved in protein-protein and protein-DNA interactions. This protein interacts with DNAJA3/hTid-1, which is a DnaJ protein reported to function as a modulator of apoptosis. Overexpression of this gene in Hela cells was shown to confer the resistance to TNF-alpha induced apoptosis, suggesting an anti-apoptotic function of this protein. This protein can be cleaved by caspase-3 during the induction of apoptosis. Alternatively spliced transcript variants encoding distinct isoforms have been reported.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RNF34
Applications ELISA, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.62 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene RNF34
Gene Accession No. Q969K3
Gene Alias AW061037; AW536122; BC004042; C88279; CARP1; CARP-1; Caspase regulator CARP1; caspases-8 and -10-associated RING finger protein 1; E3 ubiquitin-protein ligase RNF34; FLJ21786; FYVE-RING finger protein MOMO; hRFI; Human RING finger homologous to inhibitor of apoptosis protein; Momo; phafin 1; phafin-1; RFI; RIF; RIFF; ring finger protein 34; ring finger protein 34, E3 ubiquitin protein ligase; RING finger protein MOMO; RING finger protein RIFF; RING-type E3 ubiquitin transferase RNF34; RNF34
Gene Symbols RNF34
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-220 of human RNF34 (NP_0794022).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 80196
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.