Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RNF34 Polyclonal Antibody

Supplier:  Thermo Scientific PA5113296

Catalog No. PIPA5113296


Only null left
Add to Cart

Description

Description

TRUSTED_SUSTAINABILITY
Specifications

Specifications

RNF34
Polyclonal
Unconjugated
RNF34
AW061037; AW536122; BC004042; C88279; CARP1; CARP-1; Caspase regulator CARP1; caspases-8 and -10-associated RING finger protein 1; E3 ubiquitin-protein ligase RNF34; FLJ21786; FYVE-RING finger protein MOMO; hRFI; Human RING finger homologous to inhibitor of apoptosis protein; Momo; phafin 1; phafin-1; RFI; RIF; RIFF; ring finger protein 34; ring finger protein 34, E3 ubiquitin protein ligase; RING finger protein MOMO; RING finger protein RIFF; RING-type E3 ubiquitin transferase RNF34; RNF34
Rabbit
Antigen Affinity Chromatography
RUO
80196
-20°C or -80°C if preferred
Liquid
ELISA, Immunohistochemistry (Paraffin), Western Blot
0.18 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q969K3
RNF34
Recombinant Human E3 ubiquitin-protein ligase RNF34 protein (1-220AA).
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

WARNING: Cancer - www.P65Warnings.ca.gov
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.