Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ RNF36 Polyclonal Antibody
Goat Polyclonal Antibody
Supplier: Invitrogen™ PA5142046
This item is not returnable.
View return policy
Description
Additional Information: This antibody is expected to recognize both reported isoforms (NP_892030.3; NP_542783.2). This antibody is tested in Peptide ELISA: antibody detection limit dilution 1:32,000.
This gene encodes a member of the RING-B-box-coiled-coil (RBCC) family and encodes a protein with an N-terminal RING finger motif, a PRY domain and a C-terminal SPRY domain. The mouse ortholog of this gene is specifically expressed in germ cells at the round spermatid stages during spermatogenesis and, when overexpressed, induces apoptosis. Alternatively spliced transcript variants encoding distinct isoforms have been described.
Specifications
RNF36 | |
Polyclonal | |
Unconjugated | |
TRIM69 | |
4921519C19Rik; E3 ubiquitin-protein ligase TRIM69; hCG_39321; HSD34; HSD-34; RFP-like (B30.2) domain protein; RFP-like domain-containing protein trimless; RING finger B-box coiled-coil transcription factor; ring finger protein 36; RING-type E3 ubiquitin transferase TRIM69; RNF36; testis-specific RING finger protein; Trif; Trim69; Trimless; tripartite motif containing 69; tripartite motif-containing 69; tripartite motif-containing protein 69 | |
Goat | |
Ammonium Sulfate Precipitation | |
RUO | |
140691, 70928 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.5 mg/mL | |
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
Q80X56, Q86WT6 | |
TRIM69 | |
Peptide with sequence C-GENKEPLHILHPQ (aa 488-500). | |
100 μg | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction