Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ RNF36 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA556006
Description
Immunogen sequence: GDYVEMNDSI THLPSKVVIQ DITMELHCPL CNDWFRDPLM LSCGHNFCEA CIQDFWRLQA KETFCPECKM LCQYNNCTFN PVLDKLVEKI KKLPLLKGHP QCPEHGENLK LFSKPDGKLI CFQCKDARLS VGQSKEFLQI SDAV Highest antigen sequence identity to the following orthologs: Mouse - 83%, Rat - 86%.
This gene encodes a member of the RING-B-box-coiled-coil (RBCC) family and encodes a protein with an N-terminal RING finger motif, a PRY domain and a C-terminal SPRY domain. The mouse ortholog of this gene is specifically expressed in germ cells at the round spermatid stages during spermatogenesis and, when overexpressed, induces apoptosis. Alternatively spliced transcript variants encoding distinct isoforms have been described.
Specifications
RNF36 | |
Polyclonal | |
Unconjugated | |
TRIM69 | |
4921519C19Rik; E3 ubiquitin-protein ligase TRIM69; hCG_39321; HSD34; HSD-34; RFP-like (B30.2) domain protein; RFP-like domain-containing protein trimless; RING finger B-box coiled-coil transcription factor; ring finger protein 36; RING-type E3 ubiquitin transferase TRIM69; RNF36; testis-specific RING finger protein; Trif; Trim69; Trimless; tripartite motif containing 69; tripartite motif-containing 69; tripartite motif-containing protein 69 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
140691 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q86WT6 | |
TRIM69 | |
Recombinant protein corresponding to Human RNF36. Recombinant protein control fragment (Product #RP-102305). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction