Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RNF36 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA512215
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA512215 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA512215 Supplier Invitrogen™ Supplier No. PA512215
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

RNF36 Polyclonal Antibody for Western Blot

This gene encodes a member of the RING-B-box-coiled-coil (RBCC) family and encodes a protein with an N-terminal RING finger motif, a PRY domain and a C-terminal SPRY domain. The mouse ortholog of this gene is specifically expressed in germ cells at the round spermatid stages during spermatogenesis and, when overexpressed, induces apoptosis. Alternatively spliced transcript variants encoding distinct isoforms have been described.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RNF36
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene TRIM69
Gene Accession No. Q86WT6
Gene Alias 4921519C19Rik; E3 ubiquitin-protein ligase TRIM69; hCG_39321; HSD34; HSD-34; RFP-like (B30.2) domain protein; RFP-like domain-containing protein trimless; RING finger B-box coiled-coil transcription factor; ring finger protein 36; RING-type E3 ubiquitin transferase TRIM69; RNF36; testis-specific RING finger protein; Trif; Trim69; Trimless; tripartite motif containing 69; tripartite motif-containing 69; tripartite motif-containing protein 69
Gene Symbols TRIM69
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 336-365 amino acids from the central region of human RNF36 (TRIM69).
Purification Method Ammonium sulfate precipitation, Size-exclusion - Dialysis
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 140691
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.